Gene Transcripts
- Gene Symbol
- RNF19B
- Entrez Gene ID
- 127544
- Gene Name
- ring finger protein 19B
- Gene Aliases
- IBRDC3, NKLAM
- Chromosome Location
- Chr. 1: 32932498 - 32965266 on Build GRCh38
- UniGene ID
- Hs.591504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127361.2 |
NP_001120833.1
|
7, 8 |
1732 |
|
NM_001300826.1 |
NP_001287755.1
|
7, 8 |
1732 |
|
NM_153341.3 |
NP_699172.2
|
7, 8 |
1735 |
|
XM_006710356.2 |
XP_006710419.1
|
7, 8 |
2188 |
|
XM_006710358.3 |
XP_006710421.1
|
7, 8 |
2188 |
|
GenBank mRNA |
AK074486.1 |
- |
7, 8 |
1035 |
|
AK131439.1 |
- |
8, 9 |
1429 |
|
BC020595.2 |
- |
3, 4 |
440 |
|
BC062374.1 |
- |
7, 8 |
1209 |
|
BC113560.1 |
- |
7, 8 |
1102 |
8322612 |
BC143687.1 |
- |
7, 8 |
1099 |
9052201 |
View Full Product Details