Gene Transcripts

Gene Symbol
PEX10
Entrez Gene ID
5192
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
NALD, PBD6A, PBD6B, RNF69
Chromosome Location
Chr. 1: 2404802 - 2412622 on Build GRCh38
UniGene ID
Hs.732228
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002617.3 NP_002608.1 2 218
NM_153818.1 NP_722540.1 2 218
XM_011541573.1 XP_011539875.1 2 269
XM_011541576.1 XP_011539878.1 2 269
GenBank mRNA AB013818.1 - 2 218
AF060502.1 - 2 181
AK124816.1 - 2 196
BC000543.2 - 2 177 3163782
BC018198.1 - 2 200 3882695
CR542077.1 - 2 149
DA439481.1 - 2 219

View Full Product Details