Gene Transcripts

Gene Symbol
PEX10
Entrez Gene ID
5192
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
NALD, PBD6A, PBD6B, RNF69
Chromosome Location
Chr. 1: 2404802 - 2412622 on Build GRCh38
UniGene ID
Hs.732228
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002617.3 NP_002608.1 2 221
NM_153818.1 NP_722540.1 2 221
XM_011541573.1 XP_011539875.1 2 272
XM_011541576.1 XP_011539878.1 2 272
GenBank mRNA AB013818.1 - 2 221
AF060502.1 - 2 184
AK124816.1 - 2 199
BC000543.2 - 2 180 3163782
BC018198.1 - 2 203 3882695
CR542077.1 - 2 152
DA439481.1 - 2 222

View Full Product Details