Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003087.1 | 3 | 552 | ||
NR_003088.1 | 3 | 552 | |||
GenBank mRNA | AF418600.1 | - | 3 | 552 | |
AF518320.1 | - | 3 | 321 | ||
AF518321.1 | - | 3 | 407 | ||
AY063514.1 | - | 3 | 432 | ||
AY063515.1 | - | 3 | 471 | ||
AY344117.1 | - | 3 | 460 | ||
BC144077.1 | - | 2 | 285 | ||
DR004370.1 | - | 3 | 563 |