Gene Transcripts

Gene Symbol
C11orf42
Entrez Gene ID
160298
Gene Name
chromosome 11 open reading frame 42
Gene Aliases
Chromosome Location
Chr. 11: 6202997 - 6226783 on Build GRCh38
UniGene ID
Hs.278221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173525.2 NP_775796.2 2 455
XM_011519926.2 XP_011518228.1 2 3026
XM_011519927.2 XP_011518229.1 2 3026
GenBank mRNA BC031612.1 - 2 455

View Full Product Details