Gene Transcripts

Gene Symbol
RIMKLA
Entrez Gene ID
284716
Gene Name
ribosomal modification protein rimK like family member A
Gene Aliases
FAM80A, NAAGS, NAAGS-II
Chromosome Location
Chr. 1: 42380797 - 42425813 on Build GRCh38
UniGene ID
Hs.420244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173642.3 NP_775913.2 5 1294
XM_006710585.3 XP_006710648.1 5 1172
XM_017001082.1 XP_016856571.1 6 1353
GenBank mRNA BC039737.1 - 5 1288

View Full Product Details