Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321027.1 | NP_001307956.1 | 8 | 1210 | |
NM_001354.5 | NP_001345.1 | 11 | 1522 | ||
NM_205845.2 | NP_995317.1 | 10 | 1422 | ||
GenBank mRNA | AB209017.1 | - | 9 | 1177 | |
AK091194.1 | - | 10 | 2776 | ||
AK290304.1 | - | 9 | 1165 | ||
AK296419.1 | - | 8 | 1059 | ||
BC007024.1 | - | 9 | 1131 | 3681970 | |
BC063574.1 | - | 10 | 1389 | 3877666 | |
S68330.1 | - | 9 | 1115 | ||
U05598.1 | - | 9 | 1132 |