Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000939.3 | NP_000930.1 | 3 | 1291 | |
NM_001035256.2 | NP_001030333.1 | 4 | 1341 | ||
NM_001319204.1 | NP_001306133.1 | 4 | 1371 | ||
NM_001319205.1 | NP_001306134.1 | 3 | 1321 | ||
GenBank mRNA | AW951218.1 | - | 74 | ||
BC065832.1 | - | 3 | 1003 | 30392189 | |
M28636.1 | - | 1 | 142 | ||
T29537.1 | - | 1 | 74 |