Gene Transcripts

Gene Symbol
CARD18
Entrez Gene ID
59082
Gene Name
caspase recruitment domain family member 18
Gene Aliases
ICEBERG, UNQ5804, pseudo-ICE
Chromosome Location
Chr. 11: 105137721 - 105139734 on Build GRCh38
UniGene ID
Hs.56279
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021571.3 NP_067546.1 1 116
GenBank mRNA AF208005.1 - 1 108
AY358231.1 - 1 120
BC131610.1 - 1 108 40115072

View Full Product Details