Gene Transcripts

Gene Symbol
MTURN
Entrez Gene ID
222166
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Gene Aliases
C7orf41, Ells1
Chromosome Location
Chr. 7: 30134936 - 30162765 on Build GRCh38
UniGene ID
Hs.200100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152793.2 NP_690006.2 2 413
XM_005249652.3 XP_005249709.1 2 356
GenBank mRNA AK096533.1 - 2 356
AK294982.1 - 2 143
AL834391.1 - 2 347
AY054121.1 - 2 209
BC062423.1 - 2 164
BI560774.1 - 2 417
DT217816.1 - 2 348

View Full Product Details