Gene Transcripts
- Gene Symbol
- AAMDC
- Entrez Gene ID
- 28971
- Gene Name
- adipogenesis associated Mth938 domain containing
- Gene Aliases
- C11orf67, CK067, PTD015
- Chromosome Location
- Chr. 11: 77821114 - 77901281 on Build GRCh38
- UniGene ID
- Hs.745040
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316957.1 |
NP_001303886.1
|
3, 4 |
312 |
|
NM_001316958.1 |
NP_001303887.1
|
3, 4 |
312 |
|
NM_001316960.1 |
NP_001303889.1
|
3, 4 |
388 |
|
NM_001316961.1 |
NP_001303890.1
|
3, 4 |
308 |
|
NM_001316962.1 |
NP_001303891.1
|
3, 4 |
312 |
|
NM_024684.3 |
NP_078960.1
|
2, 3 |
270 |
|
NR_133637.1 |
|
2, 3 |
270 |
|
XM_011544968.2 |
XP_011543270.1
|
2, 3 |
270 |
|
XM_011544969.2 |
XP_011543271.1
|
2, 3 |
263 |
|
XM_017017620.1 |
XP_016873109.1
|
2, 3 |
292 |
|
GenBank mRNA |
AF092136.1 |
- |
3, 4 |
271 |
|
AK024688.1 |
- |
2, 3 |
229 |
|
BC002752.2 |
- |
2, 3 |
222 |
3628061 |
BC016854.2 |
- |
2, 3 |
199 |
3856546 |
BI791786.1 |
- |
3, 4 |
321 |
|
BM930331.1 |
- |
3, 4 |
257 |
|
BP270361.1 |
- |
2, 3 |
270 |
|
BX646013.1 |
- |
2, 3 |
254 |
|
CN362204.1 |
- |
3, 4 |
283 |
|
CR457332.1 |
- |
1, 2 |
124 |
|
View Full Product Details