Gene Transcripts

Gene Symbol
CHMP5
Entrez Gene ID
51510
Gene Name
charged multivesicular body protein 5
Gene Aliases
C9orf83, CGI-34, HSPC177, PNAS-2, SNF7DC2, Vps60
Chromosome Location
Chr. 9: 33264879 - 33282069 on Build GRCh38
UniGene ID
Hs.635313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195536.1 NP_001182465.1 2 331
NM_016410.5 NP_057494.3 2 331
GenBank mRNA AF132968.1 - 2 162
AF161525.1 - 2 151
AK024753.1 - 2 179
AK295744.1 - 2 171
AK315455.1 - 2 180
BC006974.1 - 2 161 3996277
BC007457.1 - 2 193 4044464
BC016698.1 - 2 210 4094210
BC018603.1 - 2 172
BC020796.1 - 2 162 4134310
BC021168.1 - 2 176 3961351

View Full Product Details