Gene Transcripts

Gene Symbol
ADIPOR2
Entrez Gene ID
79602
Gene Name
adiponectin receptor 2
Gene Aliases
ACDCR2, PAQR2
Chromosome Location
Chr. 12: 1690981 - 1788679 on Build GRCh38
UniGene ID
Hs.371642
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024551.2 NP_078827.2 5 745
XM_005253789.1 XP_005253846.1 5 867
XM_006719018.2 XP_006719081.1 6 864
XM_011521024.2 XP_011519326.1 6 875
GenBank mRNA AK025085.1 - 2 273
AK127196.1 - 6 800
AK128511.1 - 5 799
AY424280.1 - 4 624
BC051858.2 - 5 745 6527208

View Full Product Details