Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000365.5 | NP_000356.1 | 6 | 655 | |
NM_001159287.1 | NP_001152759.1 | 6 | 764 | ||
NM_001258026.1 | NP_001244955.1 | 6 | 898 | ||
GenBank mRNA | AK129488.1 | - | 6 | 667 | |
AK222638.1 | - | 6 | 644 | ||
AK298809.1 | - | 6 | 899 | ||
AK300661.1 | - | 6 | 543 | ||
AK313282.1 | - | 6 | 651 | ||
BC007086.1 | - | 6 | 651 | 4244564 | |
BC007812.2 | - | 6 | 636 | 4298901 | |
BC009329.2 | - | 6 | 642 | 4126160 | |
BC011611.2 | - | 6 | 641 | 3956298 | |
BC015100.1 | - | 6 | 656 | 3829453 | |
BC017165.1 | - | 5 | 401 | ||
BC017917.1 | - | 6 | 647 | 4296431 | |
BC018899.1 | - | 1066 | |||
CR541702.1 | - | 6 | 617 | ||
EU794668.1 | - | 6 | 656 | ||
M10036.1 | - | 6 | 984 |