Gene Transcripts
- Gene Symbol
- ZDHHC17
- Entrez Gene ID
- 23390
- Gene Name
- zinc finger DHHC-type containing 17
- Gene Aliases
- HIP14, HIP3, HSPC294, HYPH
- Chromosome Location
- Chr. 12: 76764074 - 76853701 on Build GRCh38
- UniGene ID
- Hs.4014
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015336.2 |
NP_056151.2
|
2, 3 |
360 |
|
XM_005268750.4 |
XP_005268807.1
|
2, 3 |
200 |
|
XM_011538083.1 |
XP_011536385.1
|
2, 3 |
224 |
|
XM_017019091.1 |
XP_016874580.1
|
2, 3 |
200 |
|
XM_017019092.1 |
XP_016874581.1
|
1, 2 |
104 |
|
GenBank mRNA |
AB023163.1 |
- |
2, 3 |
304 |
|
AB024494.1 |
- |
2, 3 |
847 |
|
AB097013.1 |
- |
2, 3 |
180 |
|
AB097035.1 |
- |
2, 3 |
180 |
|
AF049612.1 |
- |
2, 3 |
193 |
|
AF161412.1 |
- |
2, 3 |
192 |
|
AK292866.1 |
- |
2, 3 |
176 |
|
BC050324.1 |
- |
2, 3 |
319 |
5266097 |
DA227374.1 |
- |
2, 3 |
360 |
|
View Full Product Details