Gene Transcripts

Gene Symbol
CAGE1
Entrez Gene ID
285782
Gene Name
cancer antigen 1
Gene Aliases
CT3, CT95, CTAG3, bA69L16.7
Chromosome Location
Chr. 6: 7326654 - 7389709 on Build GRCh38
UniGene ID
Hs.721486
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170692.1 NP_001164163.1 4 1025
NM_001170693.1 NP_001164164.1 4 1025
NM_205864.2 NP_995586.1 3 807
GenBank mRNA AF414185.1 - 3 806
AK097760.1 - 4 724
BC026194.2 - 4 710 4826784
BC033545.1 - 4 710 4826548

View Full Product Details