Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000800.4 | NP_000791.1 | 4 | 1570 | |
NM_001144892.2 | NP_001138364.1 | 4 | 1090 | ||
NM_001144934.1 | NP_001138406.1 | 5 | 1283 | ||
NM_001144935.1 | NP_001138407.1 | 4 | 1189 | ||
NM_001257205.1 | NP_001244134.1 | 4 | 1480 | ||
NM_001257206.1 | NP_001244135.1 | 4 | 1477 | ||
NM_001257207.1 | NP_001244136.1 | 5 | 1223 | ||
NM_001257208.1 | NP_001244137.1 | 4 | 1221 | ||
NM_001257209.1 | NP_001244138.1 | 4 | 1236 | ||
NM_001257210.1 | NP_001244139.1 | 4 | 1220 | ||
NM_001257211.1 | NP_001244140.1 | 4 | 1218 | ||
NM_001257212.1 | NP_001244141.1 | 4 | 1087 | ||
NM_033136.3 | NP_149127.1 | 3 | 1466 | ||
NM_033137.2 | NP_149128.1 | 2 | 924 | ||
XM_005268390.4 | XP_005268447.1 | 4 | 1160 | ||
XM_011537607.2 | XP_011535909.1 | 5 | 1280 | ||
GenBank mRNA | X51943.1 | - | 3 | 1031 |