Gene Transcripts

Gene Symbol
BCL7C
Entrez Gene ID
9274
Gene Name
BCL tumor suppressor 7C
Gene Aliases
Chromosome Location
Chr. 16: 30834041 - 30894550 on Build GRCh38
UniGene ID
Hs.658547
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286526.1 NP_001273455.1 4 651
NM_004765.3 NP_004756.2 4 651
XM_011545980.2 XP_011544282.1 4 899
XM_017023885.1 XP_016879374.1 4 892
XM_017023886.1 XP_016879375.1 4 897
XM_017023887.1 XP_016879376.1 4 897
XM_017023888.1 XP_016879377.1 4 897
GenBank mRNA AJ223980.1 - 4 293
BC019071.1 - 4 427 4909589
BC058863.1 - 4 651 5167136

View Full Product Details