Gene Transcripts

Gene Symbol
APOBEC3C
Entrez Gene ID
27350
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3C
Gene Aliases
A3C, APOBEC1L, ARDC2, ARDC4, ARP5, PBI, bK150C2.3
Chromosome Location
Chr. 22: 39014260 - 39018838 on Build GRCh38
UniGene ID
Hs.441124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014508.2 NP_055323.2 3 366
GenBank mRNA AB208979.1 - 3 331
AF165520.1 - 3 370
AK094570.1 - 3 1680
AK313272.1 - 3 370
BC011739.2 - 3 366 4331363
BC021080.1 - 3 353
CR456394.1 - 3 344
KC329528.1 - 3 263

View Full Product Details