Gene Transcripts

Gene Symbol
C2orf40
Entrez Gene ID
84417
Gene Name
chromosome 2 open reading frame 40
Gene Aliases
ECRG4
Chromosome Location
Chr. 2: 106063290 - 106078155 on Build GRCh38
UniGene ID
Hs.43125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032411.2 NP_115787.1 2 221
XM_006712799.3 XP_006712862.1 3 240
XM_011511993.2 XP_011510295.1 2 158
XM_017005106.1 XP_016860595.1 2 140
GenBank mRNA AF325503.1 - 2 221
AY358601.1 - 2 182
BC021742.1 - 2 225 5260075

View Full Product Details