Gene Transcripts
- Gene Symbol
- CCDC12
- Entrez Gene ID
- 151903
- Gene Name
- coiled-coil domain containing 12
- Gene Aliases
- Chromosome Location
- Chr. 3: 46917905 - 46982031 on Build GRCh38
- UniGene ID
- Hs.631918
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001277074.1 |
NP_001264003.1
|
2, 3 |
202 |
|
NM_144716.5 |
NP_653317.2
|
3, 4 |
357 |
|
NR_102269.1 |
|
2, 3 |
220 |
|
XM_011533390.2 |
XP_011531692.1
|
2, 3 |
297 |
|
XM_011533391.2 |
XP_011531693.1
|
3, 4 |
363 |
|
XM_011533392.2 |
XP_011531694.1
|
3, 4 |
315 |
|
XM_011533394.2 |
XP_011531696.1
|
2, 3 |
839 |
|
XM_011533396.2 |
XP_011531698.1
|
2, 3 |
556 |
|
GenBank mRNA |
AK096749.1 |
- |
2, 3 |
180 |
|
AK295532.1 |
- |
2, 3 |
180 |
|
AK303160.1 |
- |
2, 3 |
194 |
|
BC020830.1 |
- |
2, 3 |
183 |
4771328 |
BM803652.1 |
- |
2, 3 |
127 |
|
CN430023.1 |
- |
3, 4 |
312 |
|
CV574708.1 |
- |
4, 5 |
433 |
|
DA952452.1 |
- |
|
220 |
|
DC379144.1 |
- |
2, 3 |
205 |
|
View Full Product Details