Gene Transcripts

Gene Symbol
BLOC1S2
Entrez Gene ID
282991
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
BLOS2, BORCS2, CEAP, CEAP11
Chromosome Location
Chr. 10: 100273278 - 100286712 on Build GRCh38
UniGene ID
Hs.34906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001342.2 NP_001001342.1 3 410
NM_001282436.1 NP_001269365.1 5 475
NM_001282437.1 NP_001269366.1 4 486
NM_001282438.1 NP_001269367.1 4 337
NM_001282439.1 NP_001269368.1 4 367
NM_173809.4 NP_776170.2 4 367
XM_017016102.1 XP_016871591.1 4 466
GenBank mRNA AB593084.1 - 4 329
AK054697.1 - 3 403
AK298969.1 - 4 335
AY278457.1 - 3 365
AY531265.1 - 4 314
BC020494.1 - 3 238 3900723
BF666010.1 - 5 444
BG545838.1 - 3 405
BG679165.1 - 4 339
BI753469.1 - 4 327
CN340962.1 - 4 454
DB456773.1 - 4 335
HY107885.1 - 3 412
HY165508.1 - 4 338

View Full Product Details