Gene Transcripts

Gene Symbol
CMTM5
Entrez Gene ID
116173
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
CKLFSF5
Chromosome Location
Chr. 14: 23376439 - 23379772 on Build GRCh38
UniGene ID
Hs.99272
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037288.1 NP_001032365.1 2 650
NM_001288746.1 NP_001275675.1 2 650
NM_138460.2 NP_612469.1 2 650
XM_017020953.1 XP_016876442.1 3 636
XM_017020954.1 XP_016876443.1 3 513
GenBank mRNA AF479262.2 - 2 651
AF527413.1 - 2 433
AK094840.1 - 1 1382
AY820135.1 - 2 206
BC013109.1 - 2 395 4214683

View Full Product Details