Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003370.2 | 11, 12 | 1135 | ||
GenBank mRNA | AK126166.1 | - | 5, 6 | 3213 | |
AK291749.1 | - | 5, 6 | 765 | ||
BC006441.1 | - | 5, 6 | 2134 | ||
BC112115.1 | - | 5, 6 | 1263 | ||
BC112117.1 | - | 5, 6 | 1263 | ||
BC143685.1 | - | 5, 6 | 1251 |