Gene Transcripts

Gene Symbol
Aqp2
Entrez Gene ID
25386
Gene Name
aquaporin 2 (collecting duct)
Gene Aliases
AQP-2, aquaporin-2
Chromosome Location
Chr. 7: 141237802 - 141242837 on Build Rnor_6.0
UniGene ID
Rn.90076
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012909.2 NP_037041.2 1 91
GenBank mRNA BC128705.1 - 1 77
D13906.1 - 1 91
L28112.1 - 1 130

View Full Product Details