Gene Transcripts
- Gene Symbol
- Akr1b8
- Entrez Gene ID
- 286921
- Gene Name
- aldo-keto reductase family 1, member B8
- Gene Aliases
- Chromosome Location
- Chr. 4: 61772064 - 61784637 on Build Rnor_6.0
- UniGene ID
- Rn.23676
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173136.1 |
NP_775159.1
|
1, 2 |
116 |
|
GenBank mRNA |
AJ277957.1 |
- |
1, 2 |
116 |
|
View Full Product Details