Gene Transcripts

Gene Symbol
Mbl1
Entrez Gene ID
24548
Gene Name
mannose-binding lectin (protein A) 1
Gene Aliases
Mbpa, Mlb1
Chromosome Location
Chr. 16: 18736154 - 18742193 on Build Rnor_6.0
UniGene ID
Rn.10273
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012599.2 NP_036731.2 2 152
GenBank mRNA BC088159.1 - 2 152
FQ209936.1 - 2 168
FQ218582.1 - 2 180

View Full Product Details