Gene Transcripts

Gene Symbol
Rps17
Entrez Gene ID
29286
Gene Name
ribosomal protein S17
Gene Aliases
Chromosome Location
Chr. 1: 143167329 - 143169915 on Build Rnor_6.0
UniGene ID
Rn.6920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017152.3 NP_058848.2 3 236
GenBank mRNA BC058484.1 - 3 240
FQ209589.1 - 3 233
FQ210096.1 - 3 226
FQ221123.1 - 3 231
FQ221680.1 - 3 226
FQ222308.1 - 3 226
FQ222382.1 - 3 238
FQ222858.1 - 3 231
FQ223398.1 - 3 233
FQ224175.1 - 3 226
FQ228331.1 - 3 231
FQ228517.1 - 3 233
FQ229910.1 - 3 233
K02933.1 - 3 234

Gene Symbol
LOC100365810
Entrez Gene ID
100365810
Gene Name
40S ribosomal protein S17-like
Gene Aliases
Chromosome Location
Chr. 8: 105087958 - 105088423 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006243604.2 XP_006243666.1 1 232

Gene Symbol
RGD1561635
Entrez Gene ID
364023
Gene Name
similar to 40S ribosomal protein S17
Gene Aliases
Chromosome Location
Chr. 13: 73150792 - 73151353 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769692.1 XP_008767914.1 1 210

View Full Product Details