Gene Transcripts

Gene Symbol
Pla2g10
Entrez Gene ID
29359
Gene Name
phospholipase A2, group X
Gene Aliases
PLA2GX, sPLA2-X
Chromosome Location
Chr. 10: 1509732 - 1520970 on Build Rnor_6.0
UniGene ID
Rn.6828
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017176.2 NP_058872.1 2 157
GenBank mRNA AF166100.1 - 2 159
AY651029.1 - 2 153

View Full Product Details