Gene Transcripts

Gene Symbol
Vmn2r55
Entrez Gene ID
100042499
Gene Name
vomeronasal 2, receptor 55
Gene Aliases
Chromosome Location
Chr. 7: 12651706 - 12684991 on Build GRCm38
UniGene ID
Mm.469870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104645.1 NP_001098115.1 3, 4 1321

View Full Product Details