Gene Transcripts
- Gene Symbol
- Psma1
- Entrez Gene ID
- 29668
- Gene Name
- proteasome (prosome, macropain) subunit, alpha type 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 183733567 - 183744586 on Build Rnor_6.0
- UniGene ID
- Rn.2668
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017278.1 |
NP_058974.1
|
8, 9 |
705 |
|
GenBank mRNA |
BC062233.1 |
- |
8, 9 |
702 |
|
D90265.1 |
- |
8, 9 |
705 |
|
FQ212620.1 |
- |
8, 9 |
674 |
|
FQ213606.1 |
- |
8, 9 |
695 |
|
FQ215179.1 |
- |
8, 9 |
684 |
|
FQ215378.1 |
- |
8, 9 |
688 |
|
FQ216107.1 |
- |
8, 9 |
698 |
|
FQ218243.1 |
- |
|
737 |
|
FQ220925.1 |
- |
8, 9 |
670 |
|
FQ222787.1 |
- |
8, 9 |
698 |
|
FQ223714.1 |
- |
8, 9 |
674 |
|
FQ225726.1 |
- |
8, 9 |
656 |
|
FQ229288.1 |
- |
8, 9 |
732 |
|
M29859.1 |
- |
8, 9 |
705 |
|
View Full Product Details