Gene Transcripts

Gene Symbol
Vmn2r95
Entrez Gene ID
328759
Gene Name
vomeronasal 2, receptor 95
Gene Aliases
Chromosome Location
Chr. 17: 18424104 - 18452324 on Build GRCm38
UniGene ID
Mm.469839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102581.1 NP_001096051.1 5 1627

View Full Product Details