Gene Transcripts

Gene Symbol
Vmn1r7
Entrez Gene ID
434016
Gene Name
vomeronasal 1 receptor 7
Gene Aliases
EG434016, Gm5568, V1rc31
Chromosome Location
Chr. 6: 57024338 - 57025273 on Build GRCm38
UniGene ID
Mm.480245
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166710.1 NP_001160182.1 1 420

View Full Product Details