Gene Transcripts

Gene Symbol
Vmn2r87
Entrez Gene ID
625131
Gene Name
vomeronasal 2, receptor 87
Gene Aliases
EG625131
Chromosome Location
Chr. 10: 130471821 - 130497379 on Build GRCm38
UniGene ID
Mm.469844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103366.1 NP_001096836.1 2 356

View Full Product Details