Gene Transcripts

Gene Symbol
Vmn2r3
Entrez Gene ID
637004
Gene Name
vomeronasal 2, receptor 3
Gene Aliases
EG637004
Chromosome Location
Chr. 3: 64258934 - 64287417 on Build GRCm38
UniGene ID
Mm.469887
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104614.1 NP_001098084.1 2 357

View Full Product Details