Gene Transcripts

Gene Symbol
Lsm6
Entrez Gene ID
78651
Gene Name
LSM6 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
1500031N17Rik, 2410088K19Rik, AI747288
Chromosome Location
Chr. 8: 78804868 - 78821152 on Build GRCm38
UniGene ID
Mm.28694
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191004.1 NP_001177933.1 5 2828
NM_030145.3 NP_084421.1 4 2729
GenBank mRNA AK141884.1 - 5 2775

View Full Product Details