Gene Transcripts

Gene Symbol
Avl9
Entrez Gene ID
78937
Gene Name
AVL9 homolog (S. cerevisiase)
Gene Aliases
5830411G16Rik, D730049P16Rik, mKIAA0241
Chromosome Location
Chr. 6: 56714905 - 56761912 on Build GRCm38
UniGene ID
Mm.105343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030235.1 NP_084511.1 16 3697
GenBank mRNA AK083605.1 - 16 3654
AK122228.1 - 16 3675
AK147365.1 - 16 3703

View Full Product Details