Gene Transcripts

Gene Symbol
Adap2
Entrez Gene ID
56826
Gene Name
ArfGAP with dual PH domains 2
Gene Aliases
Centa2
Chromosome Location
Chr. 10: 67494847 - 67527582 on Build Rnor_6.0
UniGene ID
Rn.203877
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020101.1 NP_064486.1 4 416
XM_008768109.1 XP_008766331.1 4 501
GenBank mRNA AJ238993.1 - 4 416

View Full Product Details