Gene Transcripts

Gene Symbol
Ppap2a
Entrez Gene ID
64369
Gene Name
phosphatidic acid phosphatase type 2A
Gene Aliases
Chromosome Location
Chr. 2: 44664085 - 44726820 on Build Rnor_6.0
UniGene ID
Rn.61687
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022538.2 NP_071983.2 2 214
GenBank mRNA BC061815.1 - 2 214
JQ013728.1 - 2 152
U90556.1 - 2 129

View Full Product Details