Gene Transcripts

Gene Symbol
Slc2a8
Entrez Gene ID
85256
Gene Name
solute carrier family 2, (facilitated glucose transporter) member 8
Gene Aliases
Glut8
Chromosome Location
Chr. 3: 12020227 - 12029918 on Build Rnor_6.0
UniGene ID
Rn.12923
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053494.2 NP_445946.2 10 1430
XM_008761779.1 XP_008760001.1 9 1297
GenBank mRNA AJ245935.1 - 10 1430

View Full Product Details