Gene Transcripts
- Gene Symbol
- Cand1
- Entrez Gene ID
- 117152
- Gene Name
- cullin-associated and neddylation-dissociated 1
- Gene Aliases
- Tip120, Tip120A
- Chromosome Location
- Chr. 7: 62124362 - 62162451 on Build Rnor_6.0
- UniGene ID
- Rn.32934
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054004.2 |
NP_446456.1
|
2 |
534 |
|
GenBank mRNA |
D87671.1 |
- |
2 |
536 |
|
View Full Product Details