Gene Transcripts

Gene Symbol
Klhl41
Entrez Gene ID
117537
Gene Name
kelch-like family member 41
Gene Aliases
Kbtbd10, Krp1, Sarcosin
Chromosome Location
Chr. 3: 55910177 - 55923303 on Build Rnor_6.0
UniGene ID
Rn.28875
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057191.1 NP_476539.1 1 126
GenBank mRNA AJ293948.1 - 1 126

View Full Product Details