Gene Transcripts
- Gene Symbol
- ADGRL1
- Entrez Gene ID
- 22859
- Gene Name
- adhesion G protein-coupled receptor L1
- Gene Aliases
- CIRL1, CL1, LEC2, LPHN1
- Chromosome Location
- Chr. 19: 14147737 - 14206245 on Build GRCh38
- UniGene ID
- Hs.94229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008701.2 |
NP_001008701.1
|
7, 8 |
1829 |
|
NM_014921.4 |
NP_055736.2
|
6, 7 |
1814 |
|
XM_005259818.3 |
XP_005259875.1
|
6, 7 |
1870 |
|
XM_011527796.2 |
XP_011526098.1
|
7, 8 |
1906 |
|
XM_011527798.2 |
XP_011526100.1
|
6, 7 |
1891 |
|
XM_011527801.2 |
XP_011526103.1
|
5, 6 |
1346 |
|
XM_017026475.1 |
XP_016881964.1
|
7, 8 |
1909 |
|
XM_017026476.1 |
XP_016881965.1
|
8, 9 |
2019 |
|
XM_017026477.1 |
XP_016881966.1
|
7, 8 |
1888 |
|
XM_017026478.1 |
XP_016881967.1
|
7, 8 |
1909 |
|
XM_017026479.1 |
XP_016881968.1
|
7, 8 |
1909 |
|
XM_017026480.1 |
XP_016881969.1
|
4, 5 |
1324 |
|
XM_017026481.1 |
XP_016881970.1
|
7, 8 |
1913 |
|
GenBank mRNA |
AB020628.1 |
- |
7, 8 |
1831 |
|
AF307079.1 |
- |
6, 7 |
1798 |
|
View Full Product Details