Gene Transcripts
- Gene Symbol
- Atp1b3
- Entrez Gene ID
- 11933
- Gene Name
- ATPase, Na+/K+ transporting, beta 3 polypeptide
- Gene Aliases
- AA409958, AI664000, AW212096
- Chromosome Location
- Chr. 9: 96332662 - 96364905 on Build GRCm38
- UniGene ID
- Mm.424
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007502.4 |
NP_031528.1
|
5, 6 |
735 |
|
GenBank mRNA |
AK032516.1 |
- |
5, 6 |
752 |
|
AK148495.1 |
- |
3, 4 |
440 |
|
AK151700.1 |
- |
5, 6 |
752 |
|
AK159532.1 |
- |
5, 6 |
752 |
|
AK161282.1 |
- |
5, 6 |
767 |
|
AK168273.1 |
- |
5, 6 |
751 |
|
AK191321.1 |
- |
2, 3 |
189 |
|
AK218119.1 |
- |
3, 4 |
332 |
|
BC079916.1 |
- |
5, 6 |
740 |
30665138 |
BC114530.1 |
- |
2, 3 |
160 |
|
U59761.1 |
- |
5, 6 |
738 |
|
View Full Product Details