Gene Transcripts

Gene Symbol
Prdm1
Entrez Gene ID
12142
Gene Name
PR domain containing 1, with ZNF domain
Gene Aliases
Blimp-1, Blimp1, PRDI-BF1, ZNFPR1A1, b2b1765Clo
Chromosome Location
Chr. 10: 44437174 - 44528560 on Build GRCm38
UniGene ID
Mm.4800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK133503.1 - 7 2613
AK149344.1 - 7 2504
BC129801.1 - 7 2623
JX154081.1 - 7 2446
U08185.1 - 8 2763

View Full Product Details