Gene Transcripts

Gene Symbol
Cav3
Entrez Gene ID
12391
Gene Name
caveolin 3
Gene Aliases
AI385751, Cav-3, M-cav
Chromosome Location
Chr. 6: 112459505 - 112472872 on Build GRCm38
UniGene ID
Mm.3924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007617.3 NP_031643.1 1, 2 429
GenBank mRNA BC024383.1 - 1, 2 439
CJ057698.1 - 1, 2 206
U36579.1 - 1, 2 243

View Full Product Details