Gene Transcripts
- Gene Symbol
- Gdf5
- Entrez Gene ID
- 14563
- Gene Name
- growth differentiation factor 5
- Gene Aliases
- BMP-14, Cdmp-1, bp, brp
- Chromosome Location
- Chr. 2: 155941023 - 155945364 on Build GRCm38
- UniGene ID
- Mm.4744
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC034546.1 |
- |
2 |
2046 |
|
CJ326163.1 |
- |
1 |
219 |
|
U08337.1 |
- |
2 |
2046 |
|
View Full Product Details