Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008113.3 | NP_032139.1 | 6 | 740 | |
GenBank mRNA | AK144321.1 | - | 6 | 592 | |
BC002032.1 | - | 6 | 737 | 3490167 | |
BC158003.1 | - | 6 | 605 | 9007312 | |
L42463.1 | - | 6 | 571 | ||
U73198.1 | - | 6 | 580 |