Gene Transcripts
- Gene Symbol
- Foxg1
- Entrez Gene ID
- 15228
- Gene Name
- forkhead box G1
- Gene Aliases
- 2900064B05Rik, BF-1, Bf1, Hfh9, Hfhbf1
- Chromosome Location
- Chr. 12: 49382883 - 49386867 on Build GRCm38
- UniGene ID
- Mm.4704
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U36760.1 |
- |
2 |
2096 |
|
View Full Product Details