Gene Transcripts

Gene Symbol
Hsd17b4
Entrez Gene ID
15488
Gene Name
hydroxysteroid (17-beta) dehydrogenase 4
Gene Aliases
17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
Chromosome Location
Chr. 18: 50128201 - 50196270 on Build GRCm38
UniGene ID
Mm.277857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008292.4 NP_032318.2 14, 15 1417
GenBank mRNA AK004866.1 - 14, 15 1417
AK088381.1 - 14, 15 1392
AK147182.1 - 14, 15 1389
AK152532.1 - 14, 15 1312
AK161655.1 - 14, 15 1312
AK166801.1 - 14, 15 1330
AK167060.1 - 14, 15 1367
AK169077.1 - 14, 15 1319
AK187517.1 - 2, 3 308
AK216937.1 - 2, 3 308
BC022175.1 - 14, 15 1290 4209394
X89998.1 - 14, 15 1278

View Full Product Details